Learn More
Thermo Scientific Chemicals Uridine, 99%
Description
Uridine plays a vital role in the glycolysis pathway of galactose. It is used as a precursor in the production of CDP-choline. It is an important nutrient and widely used as a dietary supplement. It improves brain cholinergic functions and hepatic mitochondrial function in certain liver toxins. It plays a major role in pain physiology and brain energy utilization to maintain ATP production under restricted oxygen conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Uridine plays a vital role in the glycolysis pathway of galactose. It is used as a precursor in the production of CDP-choline. It is an important nutrient and widely used as a dietary supplement. It improves brain cholinergic functions and hepatic mitochondrial function in certain liver toxins. It plays a major role in pain physiology and brain energy utilization to maintain ATP production under restricted oxygen conditions.
Solubility
Soluble in water, dimethylsulfoxide, and methanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 166°C to 169°C |
Assay Percent Range | 0.99 |
Quantity | 5 g |
Beilstein | 754904 |
Merck Index | 149877 |
Solubility Information | Soluble in water,dimethylsulfoxide,and methanol. |
Formula Weight | 244.2 |
Percent Purity | 99% |
Chemical Name or Material | Uridine |
RUO – Research Use Only