missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uridine, ∽99%, MP Biomedicals™
Shop All MP Biomedicals Products

Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
Specifications
Specifications
| Color | White |
| Assay Percent Range | ∽99% |
| Quantity | 10 g |
| Formula Weight | 244.2 |
| Physical Form | Powder |
| Chemical Name or Material | Uridine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction