Learn More
Gilac™ Urine Jar
Urine Jar is graduated, very stable even when empty, and benefits from a screw-on lid. Easy to clean.
Brand: Gilac™ G122421
Additional Details : Weight : 0.10300kg
Description
- 2 L urine Jar.
- Outstanding durability even in the intensive use cycle.
- Screw lid, bocal with graduation.
- Researched product ergonomics ensure optimum cleaning.
- Suitable for hospital use.
- Product can be disinfected in a bedpan washer.
Specifications
Urinal | |
Polyethylene | |
Reusable | |
0.5 gal. |
Transparent | |
Non-sterile | |
Yes | |
2 L |