Learn More
3M™ USDA Cattle/Swine Screw-top Bottle USDA25BPW
Screw-top bottle containing 25mL Buffered Peptone Water Broth for the 3M™ USDA Cattle/Swine Kits.
Brand: 3M™ 7000053826
Additional Details : Weight : 0.34500kg
Description
- Cattle/swine diluent for the 3M™ USDA Cattle/Swine Kits.
- Formulated and put together to be used for USDA testing.
- Meets the requirements of the US Pathogen Reduction Act.
Specifications
25 mL Buffered Peptone Water Broth | |
100/Cs. |