missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Usnic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is used as primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as primary and secondary intermediate.
Solubility
Partly soluble in water. Soluble in acetone, ethanol, chloroform and furfural.
Notes
Store at 4°C. Reactive with oxidizing agents.
It is used as primary and secondary intermediate.
Solubility
Partly soluble in water. Soluble in acetone, ethanol, chloroform and furfural.
Notes
Store at 4°C. Reactive with oxidizing agents.
Specifications
Specifications
Melting Point | 201°C to 203°C |
Quantity | 5 g |
Beilstein | 96698 |
Solubility Information | Partly soluble in water. Soluble in acetone,ethanol,chloroform and furfural. |
Formula Weight | 344.32 |
Percent Purity | 98% |
Chemical Name or Material | (+)-Usnic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'