missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55110-1-AP-150UL
This item is not returnable.
View return policy
Description
USP22 has 2 isoforms with molecular mass of 58 kDa and 60 kDa. The antibody is specific to USP22.
Histone deubiquitinating component of the transcription regulatory histone acetylation (HAT) complex SAGA. Catalyzes the deubiquitination of both histones H2A and H2B, thereby acting as a coactivator. Recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation and cell cycle progression.Specifications
USP22 | |
Polyclonal | |
Unconjugated | |
USP22 | |
Deubiquitinating enzyme 22, KIAA1063, Ubiquitin thiolesterase 22, USP22, USP3L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
216825, 23326, 303201 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.77 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5DU02, Q9UPT9 | |
Usp22 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |