missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | USP22 |
---|---|
Concentration | 0.77 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16816565
|
Proteintech
55110-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16806565
|
Proteintech
55110-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP22 has 2 isoforms with molecular mass of 58 kDa and 60 kDa. The antibody is specific to USP22.
Histone deubiquitinating component of the transcription regulatory histone acetylation (HAT) complex SAGA. Catalyzes the deubiquitination of both histones H2A and H2B, thereby acting as a coactivator. Recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation and cell cycle progression.Specifications
USP22 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q5DU02, Q9UPT9 | |
216825, 23326, 303201 | |
Peptide | |
Primary | |
-20°C | |
USP22 |
0.77 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Deubiquitinating enzyme 22, KIAA1063, Ubiquitin thiolesterase 22, USP22, USP3L | |
Usp22 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |