missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ UV Gel Scoops

Accessory for Fisherbrand multiSUB Horizontal Gel Systems.

£28.65 - £32.70

Specifications

Type UV Gel Scoop
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Description Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Description Price Quantity & Availability  
11857573
View Documents Promotion Details
Fisherbrand™
multiSUB Maxi Horizontal Gel Systems Size: 7cm
£28.65
Each
View alternative products
Save Up to 
»
Estimated Shipment: 13-08-2024
to see stock.
Add to basket
 
11843473
View Documents Promotion Details
Fisherbrand™
multiSUB Mini Horizontal Gel Systems Size: 20cm
£32.70
Each
View alternative products
Save Up to 
»
Estimated Shipment: 13-08-2024
to see stock.
Add to basket
 
Description

Description

  • Accessory for Fisherbrand multiSUB Horizontal Gel Systems
Specifications

Specifications

UV Gel Scoop
Videos
SDS
Documents

Documents

Product Certifications