Learn More
Faster™ UV Lamp with Aluminum Front Closure for FlowFAST or SmartFAST Cabinets
Factory fitted UV lamp for use with FlowFAST or SmartFAST Cabinets.
Brand: Faster™ FX0011846000
Code : NEW
Additional Details : Weight : 20.00000kg
Description
- Must be used in sync with aluminum front closure to prevent exposure
Specifications
Cabinet Lighting | |
FlowFAST V 18 |