Learn More
Fisherbrand™ UV Trays
Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems.
Brand:  Fisherbrand™ MS20-UV20
 View more versions of this product
Product Code. 11823473
  • £99.32 / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems
Specifications
Specifications
Sub-Gel Maxi Horizontal Gel Electrophoresis System
UV Tray
200mm x 200mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ UV Trays > Dimensions (L x W): 200 x 200mm