missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ UV Trays

Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems.

Brand:  Fisherbrand™ MS7-UV7

 View more versions of this product

Product Code. 11847573

  • ÂŁ45.05 / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems
Specifications

Specifications

Sub-Gel Mini Horizontal Gel Electrophoresis System
UV Tray
70mm x 70mm
Product Suggestions

Product Suggestions

Videos

Videos

  • Fisherbrand and Fisher Bioreagents - Focus on electrophoresis
    Fisherbrand provides a comprehensive range of horizontal gel systems for all your nucleic acid gel electrophoresis needs.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Fisherbrand and Fisher BioReagents Focus on electrophoresis

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Fisherbrand™ UV Trays > Dimensions (L x W): 70 x 70mm

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.