missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Faceshield 9705
23.48 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Size: 412 mm
Description
Specifications
Lens Coating | Uncoated |
Includes | Headband |
Size | 412 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'