missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Profi Ergo ENB20 Gloves

Product Code. 15396932
Click to view available options
Size: 10

A classic safety glove with an ergonomic fit. An extremely functional, high-quality, universal task compliance and hard-wearing safety glove.

  • Excellent ergonomic fit with good tactile feel
  • Cotton interlock safety glove for universal use, which provides high wearer comfort, feels pleasant against the skin, and provides optimum regulation of the glove climate
  • Palm and whole back of the hand with special NBR coating (nitrile rubber)
  • High flexibility
  • High water vapour absorption due to the cotton lining 
  • Proven high wearer acceptance
  • Excellent dry/wet/oily grip
  • Suitable for operating touch displays
  • Free of harmful substances in line with OEKO-Tex™ Standard 100
  • Certified in line with EN 388:2016 (2 1 2 1 X)
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance EN 388:2016 (2121X)
    Coated Area Back of the hand fully coated
    Coating Material Special NBR
    Color Orange
    Material Cotton
    Size 10
    For Use With (Application) Assembly, inspection, repairs, maintenance, and light to medium metal processing
    Glove Cuff Style Knit
    Model ENB20
    missing translation for 'provideContentCorrection'

    missing translation for 'productTitle'