missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Protective Eyewear I-vo 9160
12.06 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Frame Color:
Black, Gray
Blue, Gray
Blue, Orange
Lens Tint:
Brown
Clear
Gray
Description
- Wraparound style spectacle with excellent field of vision
- Straight side arms grip the side of the head not behind the ears. Provides a secure comfortable, slip free wear
- Combines hard/soft components - increased comfort around sensitive nose, brow and ear areas
- Fully adjustable side arms enables wearer to achieve individual fit for better protection and comfort
- 100% UV protection and metal free
Specifications
Lens Tint | Gray |
Product Type | Protective Eyewear I-vo 9160 |
Frame Color | Black, Gray |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'