Learn More
Uvex™ Unipur 6639 Polyurethane-Coated Assembly Gloves
Product Code. 15335662
Click to view available options
Size:
6
8
9
10
11
Unipur 6639 assembly gloves are suitable for precision mechanical work in dry to slightly damp work environments. The robust polyurethane coating provides abrasion-resistance, without compromising dexterity or tactility.
  • Polyamide gloves with polyurethane coating on the palm and fingertips
  • Coating offers a high level of abrasion resistance
  • Enhanced grip capabilities in dry, or slightly damp applications
  • Lightweight and sensitive, ideal for precision mechanical work
  • Dark colored material hides dirt and stains
TRUSTED_SUSTAINABILITY
Specifications
Size 8