Learn More
Uvex™ Wet Wipes
Brand: Uvex™ 9963000
Code : NEW
Additional Details : Weight : 14.40000kg
Specifications
8 x 5 in. | |
Lens Cleaning Towelette | |
Cleaning Safety Eyewear | |
100 | |
Not Autoclavable |
Individually wrapped | |
Polycarbonate, plastic, and glass lenses | |
Dispenser box | |
Single-use |