missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Winter Cap
Winter cap can be worn under helmets
£12.55
Specifications
Product Type | Balaclava |
---|---|
Material | Wool, Poly-acrylic |
Color | Black |
Closure Type | Elastic |
Disposable | Single-use |
Description
Specifications
Balaclava | |
Black | |
Single-use | |
Standard Length |
Wool, Poly-acrylic | |
Elastic | |
Cap with ear protection |