Learn More
UVP UVL-56 Handheld UV Lamp
Use these lamps for variety of applications including fluorescence of stained gels and sterilization. UVP™ Handheld UV Lamps are made of durable, lightweight plastic with an ergonomically designed handle.
Brand:  UVP 95-0006-03
Product Code. 11738221
  • £475.00 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Comfortable, ergonomically designed handle
  • On/off switch is located on the back of the lamp
  • Lamps are lightweight for easy handling
  • Durable plastic housing
  • Six watts
Specifications
Specifications
UV-A
365 nm
378 x 81 x 64 mm
230 V
Plastic
UVL-56 Handheld UV Lamp
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products