Learn More
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
11308453
|
UVP
95-0417-02 |
LMs-20E |
£2040.00
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||||
Description
- 8-watt models provide economical alternative to High Performance UV Transilluminators
- All models include hinged UV blocking cover with adjustable height and UVG long-life filter
Two single-intensity options
- 3UV models offer selection of longwave UV for extended viewing of gels without photonicking; midrange with increased fluorescence photodocumentation and nucleic acid visualization; shortwave for sample irradiation
- 2UV models provide longwave and midrange UV
1International model.
Order Info
2UV Dimensions (including cover): 9 1/2L x 13 1/4W x 4 3/4 in.H (24 x 33.7 x 12.1cm); 3UV Dimensions (including cover): 11L x 14W x 6 in.H (28 x 35.6 x 15.2cm)
Certifications
115V models are ETL, ETL/c certified; 230V1 models are CE certified
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
UVP 2UV and 3UV Benchtop Transilluminators