missing translation for 'onlineSavingsMsg'
Learn More

UVP Clear UV Blocking Covers

Product Code. p-2621985
Click to view available options
For Use With (Equipment):
3uv™ transilluminators
Benchtop transilluminators
Standard Format UV Transilluminators
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12317553

Brand: UVP 19011201

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

For Use With (Application) Gel electrophoresis
Color Clear
For Use With (Equipment) Standard Format UV Transilluminators
Includes Uv Blocking Cover
Quantity each
Size 18 x 13 in.
Product Type UV Blocking Cover
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.