Learn More
UVP J-Series Photovoltaic UV Intensity Meters: Model J-225
Photovoltaic J-Series UV meters for taking accurate and repeatable readings of UV light sources in either the 365nm or 254nm UV range
Brand:  UVP 97-0004-01
Product Code. 12813039
  • £777.00 / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With two scales for low and high intensity readings
  • Measures the intensity of shortwave UV (254nm). Thw sensor is sensitive within a range from 220 to 280nm with maximum sensitivity at 254nm.
  • For remote readings, a four foot (1.2m) cord connects the sensor to the meter
  • Two scales measure a wide range of intensity
  • An infrared filter assures accurate readings when measuring light sources producing infrared radiation
  • The 5× attenuation screen can be used for very high intensity lamp measurements
  • Meters are self-powered for maximum portability
All meters and sensors are individually serialized for traceability of calibration. Calibration is recommended every six months and is certified to meet UVP's published standards and is traceable to NIST. Calibrations are performed at UVP's factory.
Quality control
Specifications
Specifications
Photovoltaic UV Intensity Meter
Accurate and repeatable readings of shortwave UV
254nm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
UVP J-Series Photovoltaic UV Intensity Meters: Model J-225 > J-225, 254nm UV intensity meter