Learn More
UVP Replacement UV Light Tubes
Replacement UV light tubes for 4, 6, 15, 20 and 40 watt ultraviolet lamps and transilluminators
Brand:  UVP 34-0044-01
 View more versions of this product
Product Code. 12842989
  • £75.20 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
6 W
EL-series UV lamp, 3UV™ lamp, handheld UV lamp, mid size UV viewing cabinets, benchtop UV transilluminators
210.8 mm
302 nm
8.3 in.
1 Ea.