Learn More
UVP Replacement UV Light Tubes
Replacement UV light tubes for 4, 6, 15, 20 and 40 watt ultraviolet lamps and transilluminators
Brand:  UVP 34-0068-01
 View more versions of this product
Product Code. 12893848
  • £44.15 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Replacement UV Light Tube
365 nm
23 in.
1 Ea.
20 W
Model XX-20BLB UV bench lamp
584.2 mm