missing translation for 'onlineSavingsMsg'
Learn More

UVP UVX Radiometer: UVX-36 Sensor

Digital UVX Radiometer connects to one of the three interchangeable sensors (sold separately) to measure 254nm, 365nm or 302nm UV wavelengths

Brand:  UVP 97-0016-02

Product Code. 11871563

  • £682.00 / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The UVX's range switch provides selection from three intensity ranges: 0 to 20 mW/cm2, 0 to 2000 microW/cm2 and 0 to 200 microW/cm2
  • A port for connecting a chart recorder (not included) allows for continuous monitoring
  • Conversion rate: 2.8 readings/second
  • Display: 3.5 digit LCD
  • Accuracy: ±2%
  • Sensors Accuracy: ±5% (includes NIST standard)
  • Dimensions: 6.2L x 3.6W x 2 in.H (157 x 91 x 51mm)

UVX Radiometer: Powered by a 9-volt battery for up to 120 hours
10:1 attenuator: Extends the reading range up to 200mW/cm2

Sensors sold separately.

Quality control

Specifications

Specifications

UVX-36 Sensor
UVX Radiometer UVP97001502
365nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
UVP UVX Radiometer: UVX-36 Sensor > Sensor, 365nm, For UVX meter

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.