Learn More
UVP UVP XX-15 Exposure Stand
For use with UVP XX-15 UV Lamps
Brand:  UVP 18-0062-01
Product Code. 11799064
  • £246.00 / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • XX-15 Lamp models can be attached to the Exposure Stand for exposure of materials to ultraviolet
  • Shelf is adjustable to different levels to set distances from UV source
Specifications
Specifications
6 in.
Model XX-15 bench lamp
330.2 mm
20 in.
Exposure Stand
152.4 mm
13 in.
1 Ea.
508 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
UVP UVP XX-15 Exposure Stand > Stand Exposure XX-15 Lamp