missing translation for 'onlineSavingsMsg'
Learn More

BUCHI V-300 Vacuum Pump

A quiet and chemically resistant PTFE diaphragm pump. It is optimally suited to work with the I-300 / I-300 Pro Controllers for Rotavapors and parallel evaporators SyncorePlus and Multivapor.

Brand:  BUCHI 11V300200

4010.00 GBP valid until 2024-12-31
Use promo code "21962" to get your promotional price.


 View more versions of this product

Product Code. 15561280

  • £5045.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Quiet operation thanks to speed regulation
  • Unattended operation in combination with Interface I-300 / I-300 Pro when combined with optional level sensor for secondary condenser that alerts potential overflow of the receiving flask
  • High chemical resistance due to the use of high quality materials
  • Increased lifetime thanks to speed-controlled operation in conjunction with the optional Interface I-300 / I-300 Pro
  • Easy visual detection of condensate and particles due to transparent front panel
  • Seamless plug and play integration into:
    • A speed-controlled vacuum system with Interface I-300 / I-300 Pro
    • A fully automated BUCHI Rotavapor R-300 system including all process parameters

Certifications

CE/CSA/CUS

Services

1 Year

Specifications

Specifications

5 mbar
1.8 m³/h
IP20
2 bar
Vacuum Pump V-300, Woulff bottle, I-300 interface
Diaphragm Pump
Evacuating distillation apparatus, vacuum filtration, vacuum-drying cabinets, drying kilns
I-300 / I-300 Pro and R-300
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers