Learn More
Asecos™ V-MOVE-90 Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas with electronic open/close doors.
Brand:  Asecos™ 3508504035100
Additional Details : Weight : 50.00000kg
 View more versions of this product
Product Code. 15916371
  • £4288.23 / Each
Estimated Shipment: 05-11-2024
to see stock.
Description
Description
  • Optimum integration and use of space – with a depth of 86 cm, V-LINE cabinets now use the space next to laboratory fume cupboards and work benches that conventional hazardous material cabinets have left unused
  • Fire resistance of 90 minutes
  • Maximum comfort – vertical drawer technology allows access and visibility to all stored containers from both sides
  • At the push of a button the vertical drawers open and close automatically
  • Drawers glide softly and quietly out of and into the cabinet body without additional physical efforts
  • The stop function of the vertical drawers detects resistances and eliminates the risk of being trapped
  • No unauthorized use – doors lockable with cylinder lock and cabinets can be integrated by the customer into an existing master key system
  • Locking status indicator (red/green) provided
  • Always closed – serial automatic door closing after approx. 60 seconds with acoustic/visual signal before closing
  • Possible interior equipment includes height-adjustable shelves, bottom collecting sumps and guard rail systems
  • In accordance with EN 14470-1 and TRGS 510
  • Robust construction and longevity – scratch-resistant outer body with powder-coated surface
  • Ventilation – integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Specifications
860 x 449 x 1966 mm
775 x 331 x 1750 mm
4 x 50 kg sheet steel shelves; 1 x 50 kg sheet steel insert; 1 x 13 L sheet steel sump
Storage of flammable hazardous substances
Auto-close
EN 14470-1, TRGS 510
394.5 kg
Yellow
Safety storage cabinet
4 x Shelves: 50 kg each, Insert: 50 kg, Sump: 13 L, Maximum load: 120 kg
1