Learn More
Buerkle™ V2A Stainless Steel Scoop
Use this Buerkle™ V2A Stainless Steel Scoop as a measuring scoop in the laboratory. It is also suitable for use with foodstuffs
Brand:  Buerkle™ 5370-2000
46.15 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10568483
  • £72.10 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Stainless steel V2A scoop for sampling.
  • Stainless steel V2A (1.4301)
  • Sterilizable
  • Robust
Specifications
Specifications
Measuring Scoop
Autoclavable
Non-sterile
Suitable for Use with Foodstuffs
Stainless Steel (V2A)
2000 mL
AISI 304, V2A (1.4301)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ V2A Stainless Steel Scoop > Capacity: 2000mL; Diameter: 150mm