missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ V4A Stainless Steel Novartos Uno Pharma Zone Samplers (Triple-Sample)

For Unit-Dose-Sampling. It is now possible to withdraw a triple-sample without inserting the lance repeatedly into the bulk material.

Brand:  Buerkle™ 5358-1030

1681.35 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 17240584

  • £1900.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The sample inserts fit precisely and can easily be replaced
  • Inserts are available in volumes from 0 to 4.0mL
  • The insertion depths can easily be read from the laser-engraved depth scale
  • Stainless steel AISI 316L (1.4404) / AISI 316Ti (1.4571)
  • Groove-free and hand-polished
  • 25 mm Ø
Specifications

Specifications

Powder Sampler
Autoclavable
1000 mm
Stainless Steel
Reusable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Please order inserts separately!