Learn More
Buerkle™ V4A Stainless Steel PharmaPicker
The V4A Stainless Steel PharmaPicker is for sampling very small quantities of highly potent, expensive or active powders for analysis.
Brand:  Buerkle™ 5346-1000
 View more versions of this product
Product Code. 10274524
  • £586.00 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For sampling very small quantities of highly potent, expensive or active powders for analysis. The smallest sample quantities from 0.1 ml to 2.5 ml can be collected with the PharmaPicker. By simply exchanging the volume tip in the collection cylinder for 0.1–1.2 ml or 1.25–2.5 ml, the sampling volume is modified.
Specifications
Specifications
PharmaPicker
Autoclavable
T-shape
Electro-polished
60 cm
Stainless Steel (V4A)
Collection Cylinder
0.1 to 1.2 mL
Small Quantities of Powder