missing translation for 'onlineSavingsMsg'
Learn More
Learn More
V5 Epitope Tag Antibody, Agarose, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP1-30442
Additional Details : Weight : 0.00970kg
Description
V5 Epitope Tag Polyclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Immunohistochemistry, Immunoprecipitation.Specifications
V5 Epitope Tag | |
Polyclonal | |
Immunohistochemistry, Immunoprecipitation 1:10-1:500 | |
V5 epitope tag | |
Rabbits were immunized with a synthetic peptide representing amino acid residues 95 to 108 of RNA polymerase alpha subunit of simion virus 5 conjugated to KLH. | |
0.1 mg | |
Epitope Tags, V5 Epitope Tags | |
Store at 4C. Do not freeze. | |
Epitope Tag |
Immunohistochemistry, Immunoprecipitation | |
Agarose | |
PBS with 0.09% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
IgG |
For Research Use Only