Learn More
Cytiva VacuCap™ 90 PF Vacuum Filtration Devices
Innovative bottle-top filters for fast vacuum filtration of 100 mL to 5 L of aqueous solutions.
£164.00 - £253.00
Specifications
Material (Membrane) Supor (Hydrophilic PES)
Pore Size 0.8/0.2 μm
Sterility Sterile
View More Specs
Products 2
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
17154371
missing translation for 'viewDocuments' Promotion Details
Cytiva
4628
90 mm
£253.00
Pack of 10
In Stock
to see stock.
Edge
Add to basket
17194371
missing translation for 'viewDocuments' Promotion Details
Cytiva
4638
60 mm
£164.00
Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Supor (Hydrophilic PES)
0.8/0.2 μm
Sterile
Videos
SDS
Documents
Documents
Certificates
Product Content Correction
Product Title
Cytiva VacuCap™ 90 PF Vacuum Filtration Devices