missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) VacuCap™ 90 Vacuum Filtration Devices
Innovative bottle-top filters for fast vacuum filtration of 100 mL to 5 L of aqueous solutions.
Brand: Cytiva (formerly Pall Lab) 4631
Additional Details : Weight : 1.34900kg
Description
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Supor (Hydrophilic PES) | |
Sterile | |
60 mm |
1 μm | |
30 cm2 |