missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One VACUETTE™ Glucose Blood Collection Tubes

Product Code. 16524500
Click to view available options
Additive:
FC Mix
Closure Color:
Pink
Capacity (Metric):
2 mL
Quantity:
1200: 50/Pk. 24 Pk.
This item is not returnable. View return policy

Product Code. 16524500

Brand: Greiner Bio-One 454510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VACUETTE™ Glucose Collection Tubes are ideal for glucose determinations. They contain sodium fluoride as stabiliser with a choice of potassium oxalate, K3EDTA or sodium heparin as anticoagulant. The VACUETTE™ FC Mix tube enables exact measurement of the in-vivo blood sugar level, which makes it particularly suitable for the diagnosis of Diabetes mellitus.

  • Tubes are supplied in packs of 50 pieces
  • Available with potassium oxalate, K3EDTA or sodium heparin as anticoagulant
  • Made out of virtually unbreakable PET plastic
  • They provide improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • CE marked
TRUSTED_SUSTAINABILITY

Specifications

Additive FC Mix
Material PET
Sterility Sterile
Closure Type Push Cap
Capacity (Metric) 2 mL
Diameter (Metric) Inner 13 mm
Closure Included Yes
Closure Color Pink
Quantity 1200: 50/Pk. 24 Pk.
For Use With (Application) Blood collection
Length (Metric) 75 mm
Description FC Mix tubes contain citrate/citrit acid buffer reducing the pH value in the sample immediately after blood collection and the actual in vivo level is stabilized right from the start. The tube is optimal for the diagnosis of diabetes mellitus and gestational diabetes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.