Learn More
Integra Biosciences™ VACUSAFE Aspiration System
Use this aspiration system for the safe disposal of biological liquids. Integra Biosciences™ VACUSAFE Aspiration System is a compact, all-in-one laboratory vacuum pump, providing a safe and efficient way to collect and contain biological liquid waste.
Brand: Integra Biosciences™ 158300
Code : 04
Additional Details : Weight : 0.10000kg
Description
- Enables Biosafety level 1, 2 and 3 labs to comply with the stringent user safety guidelines for handling contaminated solutions
- Maximizes productivity and minimizing noise and operational inconveniences
- Equipped with vacuum source protection, a shatterproof bottle and a secure stand that holds the bottle in place
- Includes integrated vacuum pump, complete with collection bottle, bottle handle, tubing set with filter, mains adapter and VACUBOY hand operator (155 510).
- 3 liter bottle (safety coated glass)
- Level detection and lid with quick tubing couplings (PVDF)
- Self-closing connectors to avoid escape of drops or aerosols
- All-in-one system includes all necessary components
- Straightforward operation – simply switch on, set the desired vacuum and start working
- Integrated pump / valve is switched off automatically when the vacuum is established in the bottle
- Noise of the integrated pump is well below 65 dB(A)
- Quick tubing connectors speed up bottle removal
Specifications
2 L Glass bottle, Lid with quick couplings and level detection, Tubing set with filter, Mains adapter: 100-240VAC, 50/60Hz, VACUBOY hand operator with 1-channel SS adapter 40 mm | |
VacuSafe Comfort is designed for aspiration, disposal, or filtration of biological /chemical fluids in a clean and professional way. | |
8 L/min. | |
-300 to -600 mbar (adjustable) |