Learn More
Integra Biosciences™ Vacusafe Tubing Set with Filter
Use this filter and tubing set for the aspiration of biological liquid waste. Integra Biosciences™ Vacusafe Tubing Set with Filter is designed for use with VACUSAFE Aspiration Systems.
Brand:  Integra Biosciences™ 158342
Code : 04
Additional Details : Weight : 0.11250kg
Product Code. 11696620
  • £66.15 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Silicone tubing set with hydrophobic filter
  • Includes: Tubing for connection of hand operator to bottle, 180 cm, silicone (158 332); tubing for connection of instrument to bottle, 70 cm, silicone (158 331); filter for protection of vacuum source, 0.45 μm, hydrophobic (158 015)
  • For use with VACUSAFE Aspiration Systems
VACUSAFE Aspiration System is a compact, all-in-one laboratory vacuum pump, providing a safe and efficient way to collect and contain biological liquid waste.
Specifications
Specifications
VACUSAFE Aspiration Systems
Aspiration of biological liquid waste
Tubing for connection of hand operator to bottle, 180 cm, silicone (Mfr. No. 158 332); tubing for connection of instrument to bottle, 70 cm, silicone (Mfr. No. 158 331); filter for protection of vacuum source, 0.45 μm, hydrophobic (Mfr. No. 158 015)