missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Vacushield™ Vent Device

Protects valves and pump components from damage due to liquids.

Brand:  Cytiva 4402

118.31 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 17154301

  • £136.00 / Pack of 3
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Allows air and gases to pass through freely while blocking aqueous solutions and aerosol contaminants
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTFE
Non-sterile
Stepped Hose Barb
3/Pk.
0.2 μm
19.6 cm2
Stepped Hose Barb
50 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Vacushield™ Vent Device > 0.2 μm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.