Learn More
BD Vacutainer™ Safety-Lok™ Blood Collection Set
Designed to help prevent needle stick injuries. BD™ Vacutainer™ Safety-Lok™ Blood Collection Set are sterile, closed systems for venous blood collection. Each versatile winged infusion set includes an irreversible safety mechanism that can be activated with one hand.
Brand:  BD 367288
 View more versions of this product
Product Code. 12927716
  • £168.00 / Pack of 200
Explore more special offers
Description
Description
Safety-Lok™ blood collection sets are sterile, closed systems for venous blood collection. They can also be used for short-term infusions of up to two hours. These winged infusion sets include a hypodermic needle, color-coded wings, an integrated safety shield, small-bore tubing and a connector. Many also include a Luer adapter.
  • Safety mechanism protects against needle injuries following successful venipuncture
  • Safety shield pushes over the needle, surrounding it completely
  • Safety mechanism engages irreversibly with an audible click over the needle
  • Single-handed activation so attention remains focused on the patient and the puncture site
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blood Collection Set
23 G
Venous blood collection, Short-term infusions
Yes
305 mm
Light Blue
Sterile
With luer adapter, integrated safety shield
19 mm
200 Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Vacutainer™ Safety-Lok™ Blood Collection Set > Gauge: 23; Tubing Length: 305mm, Wing color: Light blue; With Luer adapter