Learn More
BD Vacutainer™ Citrate Tube
Contains buffered trisodium citrate solution. BD™ Vacutainer™ Citrate Tube contains an anticoagulant additive equivalent to 3.2% trisodium citrate for coagulation investigations. It is also suitable for carrying out special test procedures, such as the platelet function test PFA-100.
Brand: BD 367691
Description
Trisodium citrate works as an anticoagulant by forming complexes with metal ions such as calcium, thereby inhibiting the coagulation cascade. Anticoagulation with trisodium citrate is reversible. The blood to additive ratio is 9:1.
BD Vacutainer™ Citrate Tubes are available in glass or Plus PET, both with blue Hemogard™ closures.
- Both tubes feature clinically equivalent performance
- Glass tubes are recognized as the global “Gold Standard” and have an internal coating of a special silicone to minimize contact activation
- Plus PET tubes include a Minimum Fill Indicator marking and feature innovative tube geometry that minimizes tube headspace and associated platelet activation (to optimize APTT monitoring of unfractionated heparin patients)
- Glass tubes are not recommended for centrifugation at more than 2200 g in a swing-out rotor (1300 g for a fixed angle rotor), and so may not be appropriate for use with platelet free plasma

Specifications
Venous Blood Sampling Coagulation Tube | |
Glass | |
4.5 mL | |
75 mm | |
Block |
Sodium Citrate | |
Hemogard | |
Light Blue | |
Used for coagulation studies | |
13 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.