missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Citrate Tube

Contains buffered trisodium citrate solution. BD™ Vacutainer™ Citrate Tube contains an anticoagulant additive equivalent to 3.2% trisodium citrate for coagulation investigations. It is also suitable for carrying out special test procedures, such as the platelet function test PFA-100.

Brand:  BD 363093

 View more versions of this product

Product Code. 12987656

  • £561.00 / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Trisodium citrate works as an anticoagulant by forming complexes with metal ions such as calcium, thereby inhibiting the coagulation cascade. Anticoagulation with trisodium citrate is reversible. The blood to additive ratio is 9:1.

BD Vacutainer™ Citrate Tubes are available in glass or Plus PET, both with blue Hemogard™ closures.

  • Both tubes feature clinically equivalent performance 
  • Glass tubes are recognized as the global “Gold Standard” and have an internal coating of a special silicone to minimize contact activation
  • Plus PET tubes include a Minimum Fill Indicator marking and feature innovative tube geometry that minimizes tube headspace and associated platelet activation (to optimize APTT monitoring of unfractionated heparin patients)
  • Glass tubes are not recommended for centrifugation at more than 2200 g in a swing-out rotor (1300 g for a fixed angle rotor), and so may not be appropriate for use with platelet free plasma
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Venous Blood Sampling Coagulation Tube
PET
1.8 mL
75 mm
Block
Sodium Citrate
Hemogard
Light Blue
Used for coagulation studies
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD Vacutainer™ Citrate Tube > Capacity: 1.8mL; Material: PET

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.