Learn More
BD Vacutainer™ Flashback Blood Collection Needles
Provides instant confirmation of needle placement through a visual flashback system. BD Vacutainer™ Flashback Blood Collection Needles are sterile, multi-sample needles with a safety valve and pre-attached BD tube holder. Available in several lengths and gauges.
£168.00 - £437.00
Specifications
Product Type Blood Collection Needle
Sterility Sterile
Connectors Vacutainer
For Use With (Application) Blood Collection
For Use With (Equipment) Complements the entire Vacutainer range of blood collection devices and accessories
View More Specs
Products 3
Product Code Brand Gauge Length (Metric) Color Price Quantity & Availability  
Product Code Brand Gauge Length (Metric) Color Price Quantity & Availability  
12927726
View Documents
BD
301746
21 G 25.4 mm Green
£437.00
Pack of 1000
12937726
View Documents
BD
301747
22 G 25.4 mm Black
£436.70
Pack of 50
12947726
View Documents
BD
301748
22 G 31.75 mm Black This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
The BD Vacutainer™ system is optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process.
  • Provides instant confirmation of needle placement through a unique visual flashback indication
  • Avoids the need to probe for a vein or to stick a patient multiple times when the vein is hard to find
  • Ensures easy training of personnel and full utilization of closed system collection
  • Multi-sample needle ideally complements the range of Vacutainer blood collection line
Specifications
Specifications
Blood Collection Needle
Vacutainer
Complements the entire Vacutainer range of blood collection devices and accessories
Sterile
Blood Collection