Learn More
BD Vacutainer™ Hemogard™ Closure Plastic K2-Edta Tube
Used to obtain a whole blood or EDTA plasma sample. Vacutainer™ Hemogard™ Closure Plastic K2-Edta Tube contains K2EDTA which is a liquid solution that is spray coated onto the interior surface of the BD Vacutainer Plus Tubes.
Brand:  BD 367873
 View more versions of this product
Product Code. 12957666
  • £445.00 / Pack of 1000
Edge
Explore more special offers
Description
Description
Lavender Hemogard™ closure
Paper label
TRUSTED_SUSTAINABILITY
Specifications
Specifications
K2 EDTA Tube
Plastic
6 mL
100 mm
13 mm
K2 EDTA
Hemogard
Lavender
Block
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Vacutainer™ Hemogard™ Closure Plastic K2-Edta Tube > Capacity: 6mL; Label Type: Block