missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Hemogard™ Closure Plastic K3-Edta Tube

Product Code. p-8000827
Click to view available options
Capacity (Metric):
2 mL
4 mL
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 12967666

Brand: BD 367836

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use with whole blood or EDTA plasma sample. BD Vacutainer™ Hemogard™ Closure Plastic K3-EDTA Tube contains the additive K3 Potassium Salt of EDTA that acts as a coagulant.

  • Lavender Hemogard™ Closure
  • Used for whole blood or EDTA plasma sample
  • The additive K3 Potassium Salt of EDTA is a liquid in the Vacutainer Tube and acts as a coagulant
  • Available in sizes 2ml, 3ml and 4ml.
  • The difference between K2EDTA and K3EDTA is that the K3 is a liquid solution in the tubes, whereas the K2 is a liquid solution that is spray coated onto the tubes
TRUSTED_SUSTAINABILITY

Specifications

Product Type K2 EDTA Tube
Additive K3 EDTA Additive
Material Plastic
Closure Type Hemogard
Capacity (Metric) 2 mL
Label Type Block
Diameter (Metric) Outer 13 mm
Certifications/Compliance ICSH (International Council Society of Haematology)
compliance-icons
Product Identifier
  • BD Vacutainer K3E 3.6 mg Blood Collection Tubes
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Tripotassium EDTA Dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.