missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Serum Tubes

Product Code. p-8008926
Click to view available options
Capacity (Metric):
2 mL
4 mL
6 mL
10 mL
Tube Size:
13 x 75 mm
13 x 100 mm
16 x 100 mm
Unit Size:
Pack of 100
Pack of 1000
This item is not returnable. View return policy

Product Code. 12927686

Brand: BD 368493

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to obtain a serum sample. BD™ Vacutainer™ Serum Tubes are made of durable PET plastic with a Hemogard™ closure. It has silica particles added to trigger coagulation, and are marked CAT and CE.

In order to obtain serum samples from plastic tubes, the tube must have a coagulation activator added. As the plastic surface alone is insufficient to trigger the coagulation within an acceptable time, BD Vacutainer™ plastic serum tubes have silica particles added for this purpose.

  • These tubes are marked with the acronym CAT (Clot Activator Tube)
  • Recommended minimum time for coagulation from patients who have not been treated with anticoagulants is 60 minutes
  • Centrifuging conditions: ≤1300 g for 10 minutes at 18–25°C
TRUSTED_SUSTAINABILITY

Specifications

Product Type Serum Tube
Additive Silica
Material PET
Closure Type Hemogard
Capacity (Metric) 2 mL
Closure Color Red
Tube Size 13 x 75 mm
For Use With (Application) Used to obtain a serum sample
Label Type Block
Certifications/Compliance CE, CAT
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.