Learn More
Fisherbrand™ Vacuum Aspiration Advantage System Accessory, Eight-channel Adapter
For use with Vacuum Aspiration Advantage System. The 8-channel adapter with ejector is made of plastic, and can be used with generic tips to provide uniform aspiration across all channels and minimise variations in sample volumes.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 1.25690kg
Description
- For use with Vacuum Aspiration Advantage System
- Enhances precision and accuracy across all channels, minimises sample volume variation
- The elector allows for easy and efficient dispensing of the samples
- Streamlines the workflow and reduces the risk of sample cross-contamination
- Compatible with a variety of sample containers, such as microplates or tubes
Specifications
Plastic | |
Adapter | |
Aspiration Advantage System |
8 | |
Aspiration of biological and medical liquids and fluids, Cell culture aspiration, Disposal of biological and medical fluids, Rinsing of sample cultures |