Learn More
Gardner Denver Welch™ Kit 9 Vacuum pump control package with MRV 100, DN 16 KF
For rotary vane pumps with KF16 flanges. Cost-effective package for controlling vacuum pressure up to 0.001mbar.
Brand:  Gardner Denver Welch™ 330053
Product Code. 12850593
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Vacuum control package consists of:
  • MRV100 vacuum controller
  • PIZA111cr-GOLD vacuum sensor
  • KF16 measuring T-piece
  • 2-way solenoid valve
  • Clamps, centring rings and adapters
  • Electrical connections
Specifications
Specifications
DN 16 KF
Vacuum Control Kit
Rotary vane pumps with KF16 flanges