Learn More
BINDER™ Vacuum Pump Connection Kit
For Use With: series VDL
Brand: BINDER™ 8012-0621
This item is not returnable.
View return policy
Description
- For vacuum pump VP 4
- Consisting of clamping rings, universal centering rings, transition piece, anti-static vacuum hose, union nut, and elbow
Specifications
Vacuum pump connection kit (ATEX) - for vacuum pump VP 4; consisting of clamping rings, universal centering rings, transition piece, anti-static vacuum hose, union nut, and elbow | |
Series VDL | |
Vacuum Pump Connection Kit |