Learn More
Heidolph™ Vacuum Pump Hei-VAC Valve Tec
Hei-VAC Valve Tec is a valve-regulated vacuum pump.
Brand: Heidolph™ 591-00160-03
Additional Details : Weight : 19.90000kg
Description
- Recommended for solvents with low or medium boiling points
- Vacuum can be controlled via valve operated or manual vacuum controllers
- A Vacuum valve is necessary if you want to combine this vacuum pump with the Hei-VAP Expert Control / Hei-VAP Ultimate Control rotary evaporators
- Condenser is not in scope of delivery
Specifications
UK Plug | |
IP20 | |
230 V |