Learn More
Heidolph™ Vacuum Pump Hei-VAC Vario Tec
Hei-VAC Vario Tec is an RPM-regulated vacuum pump.
Brand: Heidolph™ 591-00171-03
Additional Details : Weight : 19.90000kg
Description
- For Hei-VAP Expert Control / Hei-VAP Untimate Control
- Condenser is not in scope of delivery
- Recommended for solvents with low or medium boiling points
Specifications
UK Plug | |
IP20 | |
230 V |