Learn More
Heidolph™ Vacuum Pump Rotavac Valve Control
Rotavac Valve Control is a valve-regulated vacuum pump.
Brand: Heidolph™ 591-00130-03
Additional Details : Weight : 13.00000kg
Description
- Suction capacity for up to three rotary evaporators at the same time
- Vacuum can be controlled via valve operated or manual vacuum controllers
- Depending on your application you can switch on and off the vacuum pump via switchbox
- A Vacuum valve is necessary, if you want to combine this vacuum pump with the Hei-VAP Expert Control / Hei-VAP Ultimate Control rotary evaporators
- Condenser is not in scope of delivery
Specifications
UK Plug | |
IP40 | |
230 V |